Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286040.1 | NP_001272969.1 | 17 - 18 | 2748 | 93 | |
NM_001286041.1 | NP_001272970.1 | 16 - 17 | 2685 | 93 | ||
NM_181413.4 | NP_852078.1 | 17 - 18 | 2748 | 93 | ||
GenBank mRNA | AK080878.1 | - | 6 - 7 | 1383 | 93 | |
AK129094.1 | - | 16 - 17 | 2619 | 93 | ||
AK147273.1 | - | 17 - 18 | 2683 | 93 | ||
AK160806.1 | - | 4 - 5 | 422 | 93 | ||
AK169828.1 | - | 6 - 7 | 694 | 93 | ||
BC010633.1 | - | 5 - 6 | 631 | 93 | ||
BC050847.1 | - | 17 - 18 | 2714 | 6507130 | 93 | |
BC057331.1 | - | 5 - 6 | 892 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |