Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286040.1 | NP_001272969.1 | 1 - 2 | 336 | 71 | |
NM_001286041.1 | NP_001272970.1 | 1 - 2 | 336 | 71 | ||
NM_181413.4 | NP_852078.1 | 1 - 2 | 336 | 71 | ||
GenBank mRNA | AK129094.1 | - | 1 - 2 | 270 | 71 | |
AK147273.1 | - | 1 - 2 | 271 | 71 | ||
BC027740.1 | - | 1 - 2 | 322 | 71 | ||
BC050847.1 | - | 1 - 2 | 302 | 6507130 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |