Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026274.4 | NP_080550.3 | 3 - 4 | 1042 | 150 | |
GenBank mRNA | AK032416.1 | - | 3 - 4 | 1005 | 150 | |
AK076799.1 | - | 3 - 4 | 615 | 150 | ||
AK141443.1 | - | 3 - 4 | 592 | 150 | ||
AK151018.1 | - | 3 - 4 | 1012 | 150 | ||
AK152024.1 | - | 3 - 4 | 1012 | 150 | ||
AK172567.1 | - | 3 - 4 | 1008 | 150 | ||
AK173313.1 | - | 3 - 4 | 661 | 150 | ||
BC010833.1 | - | 3 - 4 | 657 | 4235165 | 150 | |
BC054121.1 | - | 3 - 4 | 1028 | 6393910 | 150 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |