Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026274.4 | NP_080550.3 | 5 - 6 | 1283 | 65 | |
GenBank mRNA | AK032416.1 | - | 5 - 6 | 1246 | 65 | |
AK076799.1 | - | 5 - 6 | 856 | 65 | ||
AK134544.1 | - | 2 - 3 | 150 | 65 | ||
AK141443.1 | - | 5 - 6 | 833 | 65 | ||
AK150941.1 | - | 4 - 5 | 758 | 65 | ||
AK151018.1 | - | 5 - 6 | 1253 | 65 | ||
AK152024.1 | - | 5 - 6 | 1253 | 65 | ||
AK152435.1 | - | 4 - 5 | 758 | 65 | ||
AK172567.1 | - | 5 - 6 | 1249 | 65 | ||
AK173313.1 | - | 5 - 6 | 902 | 65 | ||
BC010833.1 | - | 5 - 6 | 898 | 4235165 | 65 | |
BC054121.1 | - | 5 - 6 | 1269 | 6393910 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |