Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110015.1 | NP_001103485.1 | 8 - 9 | 1082 | 156 | |
NM_001110016.1 | NP_001103486.1 | 8 - 9 | 1082 | 156 | ||
NM_144863.4 | NP_659112.2 | 8 - 9 | 1082 | 156 | ||
GenBank mRNA | AK041763.1 | - | 8 - 9 | 1002 | 156 | |
AK045396.1 | - | 8 - 9 | 1055 | 156 | ||
AK140019.1 | - | 8 - 9 | 1007 | 156 | ||
AK156822.1 | - | 1083 | 156 | |||
AK171683.1 | - | 8 - 9 | 1020 | 156 | ||
AK172054.1 | - | 8 - 9 | 1007 | 156 | ||
AK172377.1 | - | 8 - 9 | 1012 | 156 | ||
BC022132.1 | - | 8 - 9 | 1054 | 156 | ||
BC025809.1 | - | 8 - 9 | 996 | 156 | ||
BC094453.1 | - | 8 - 9 | 5068941 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |