Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031524.1 | - | 9 - 10 | 3757 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |