Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173754.4 | NP_776115.2 | 12 - 13 | 2116 | 59 | |
GenBank mRNA | AK083271.1 | - | 12 - 13 | 1954 | 59 | |
BC021474.1 | - | 11 - 12 | 1406 | 5343429 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |