Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175488.6 | NP_780697.2 | 8 - 9 | 950 | 80 | |
GenBank mRNA | AK029803.1 | - | 8 - 9 | 951 | 80 | |
AK030198.1 | - | 8 - 9 | 939 | 80 | ||
AK030205.1 | - | 5 - 6 | 600 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |