Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176940.5 | NP_795914.3 | 9 - 10 | 2718 | 79 | |
GenBank mRNA | AK034868.1 | - | 2417 | 79 | ||
AK038792.1 | - | 8 - 9 | 2411 | 79 | ||
AK087622.1 | - | 9 - 10 | 2723 | 79 | ||
AK162888.1 | - | 9 - 10 | 2713 | 79 | ||
BC082552.1 | - | 9 - 10 | 2658 | 79 | ||
BC144786.1 | - | 7 - 8 | 2308 | 9053334 | 79 | |
BC144787.1 | - | 7 - 8 | 2308 | 79 | ||
BC145703.1 | - | 7 - 8 | 2308 | 40131006 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |