Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177081.3 | NP_796055.1 | 5 - 6 | 72 | ||
GenBank mRNA | AK049175.1 | - | 4 - 5 | 72 | ||
AK083305.1 | - | 5 - 6 | 72 | |||
BC098475.1 | - | 5 - 6 | 30044369 | 72 | ||
BC138194.1 | - | 4 - 5 | 522 | 8861213 | 72 | |
BC145190.1 | - | 4 - 5 | 522 | 9053741 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |