Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177092.4 | NP_796066.1 | 2 - 3 | 257 | 101 | |
GenBank mRNA | AK086975.1 | - | 2 - 3 | 224 | 101 | |
BB617432.1 | - | 2 - 3 | 292 | 101 | ||
BC096447.1 | - | 2 - 3 | 138 | 6310686 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |