Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177158.5 | NP_796132.2 | 5 - 6 | 459 | 59 | |
GenBank mRNA | AK030993.1 | - | 5 - 6 | 452 | 59 | |
BC044761.1 | - | 5 - 6 | 495 | 59 | ||
BC046530.1 | - | 5 - 6 | 495 | 5329601 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |