Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcp11l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177190.5 | NP_796164.1 | 3 - 4 | 503 | 89 | |
GenBank mRNA | AK047525.1 | - | 2 - 3 | 329 | 89 | |
AK082024.1 | - | 3 - 4 | 505 | 89 | ||
BC116926.1 | - | 2 - 3 | 324 | 40126245 | 89 | |
BC116932.1 | - | 2 - 3 | 324 | 40126251 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |