Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Svopl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177200.4 | NP_796174.2 | 9 - 10 | 898 | 117 | |
GenBank mRNA | AK034997.1 | - | 9 - 10 | 899 | 117 | |
AK137043.1 | - | 9 - 10 | 899 | 117 | ||
BC058741.1 | - | 9 - 10 | 881 | 6309979 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |