Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177448.4 | NP_803231.1 | 2 - 3 | 308 | 92 | |
GenBank mRNA | AK049560.1 | - | 2 - 3 | 271 | 92 | |
AK133854.1 | - | 2 - 3 | 316 | 92 | ||
AY157609.1 | - | 2 - 3 | 265 | 92 | ||
BC052831.1 | - | 2 - 3 | 312 | 30041797 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |