Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177465.4 | NP_803416.2 | 12 - 13 | 2529 | 57 | |
GenBank mRNA | AK040446.1 | - | 4 - 5 | 1030 | 57 | |
AY771616.1 | - | 13 - 14 | 2446 | 57 | ||
AY771617.1 | - | 12 - 13 | 2533 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |