Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177732.4 | NP_808400.1 | 5 - 6 | 478 | 122 | |
GenBank mRNA | AK049180.1 | - | 5 - 6 | 479 | 122 | |
AK049462.1 | - | 5 - 6 | 398 | 122 | ||
AK052968.1 | - | 5 - 6 | 398 | 122 | ||
AK140305.1 | - | 5 - 6 | 770 | 122 | ||
AK164517.1 | - | 5 - 6 | 398 | 122 | ||
AK169775.1 | - | 5 - 6 | 380 | 122 | ||
AK220330.1 | - | 5 - 6 | 388 | 122 | ||
BC115984.1 | - | 6 - 7 | 379 | 40105299 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |