Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177750.6 | NP_808418.1 | 4 - 5 | 530 | 79 | |
GenBank mRNA | AK047241.1 | - | 2 - 3 | 2250 | 79 | |
BC117734.1 | - | 2 - 3 | 186 | 40089821 | 79 | |
BC117735.1 | - | 2 - 3 | 186 | 40089822 | 79 | |
BC129957.1 | - | 2 - 3 | 186 | 40089827 | 79 | |
BC129958.1 | - | 2 - 3 | 186 | 40089829 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |