Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301173.1 | NP_001288102.1 | 1 - 2 | 48 | 68 | |
NM_177791.3 | NP_808459.1 | 1 - 2 | 48 | 68 | ||
GenBank mRNA | AK076644.1 | - | 1 - 2 | 40 | 68 | |
BC092530.1 | - | 1 - 2 | 48 | 6744085 | 68 | |
BC125431.1 | - | 1 - 2 | 40 | 40129946 | 68 | |
BC125433.1 | - | 1 - 2 | 40 | 40129948 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |