Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177815.3 | NP_808483.2 | 3 - 4 | 299 | 67 | |
GenBank mRNA | AK086390.1 | - | 3 - 4 | 288 | 67 | |
BB652085.1 | - | 4 - 5 | 443 | 67 | ||
BC082539.1 | - | 3 - 4 | 267 | 67 | ||
BC094336.1 | - | 3 - 4 | 267 | 67 | ||
BC116368.2 | - | 3 - 4 | 280 | 67 | ||
BC116369.2 | - | 3 - 4 | 280 | 67 | ||
BC141275.1 | - | 3 - 4 | 293 | 9055887 | 67 | |
BC145606.1 | - | 3 - 4 | 293 | 9054172 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |