Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285963.1 | NP_001272892.1 | 4 - 5 | 729 | 148 | |
NM_001285965.1 | NP_001272894.1 | 4 - 5 | 729 | 148 | ||
GenBank mRNA | AK040156.1 | - | 3 - 4 | 718 | 148 | |
EF494892.1 | - | 4 - 5 | 729 | 148 | ||
EU099299.1 | - | 4 - 5 | 729 | 148 | ||
EU099300.1 | - | 4 - 5 | 729 | 148 | ||
EU650234.1 | - | 3 - 4 | 703 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |