Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177910.3 | NP_808578.1 | 3 - 4 | 489 | 76 | |
GenBank mRNA | AK088295.1 | - | 3 - 4 | 471 | 76 | |
AK148125.1 | - | 489 | 76 | |||
AK158470.1 | - | 3 - 4 | 489 | 76 | ||
AK165816.1 | - | 3 - 4 | 491 | 76 | ||
BC061207.1 | - | 4 - 5 | 363 | 30249330 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |