Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454874 |
AK153812.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454874 |
BC055377.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454874 |
M90388.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
454874 |
NM_008979.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
318747 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
84129 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168549 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168549 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
AK149459.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
AY575783.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
BC132100.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
NM_177920.5 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P162955 |
AK050361.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P112189 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P124273 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168549 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15116316 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15116329 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15116340 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15116324 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15116335 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15116308 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692816 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692818 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692767 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692804 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692757 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692796 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692764 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692774 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692754 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692801 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692812 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692807 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692798 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692779 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692786 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692810 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692763 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32692777 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692774 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692750 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15116322 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692725 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692732 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692771 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692799 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692751 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692794 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692813 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692782 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692773 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692809 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692743 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692786 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692733 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692804 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692748 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692812 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692747 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692807 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692745 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692767 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692787 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692765 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692736 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692798 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692796 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692758 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692790 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692778 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692739 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692729 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692779 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14667256 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692761 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692738 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692735 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692744 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692801 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692757 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692731 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692727 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692805 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692811 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15116332 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14908705 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692769 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692754 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692734 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692793 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692722 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692766 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692775 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692780 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692753 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32692788 |
- |
- |