Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
NM_001039376.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
NM_001039376.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
NM_001289701.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
NM_001289702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
NM_178080.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
NM_178080.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
NR_110360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019882.mm.1 |
TC0300002543.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019880.mm.1 |
NR_110360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019880.mm.1 |
TC0300002543.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
NM_001039376.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
NM_001039376.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
NM_001289701.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
NM_001289702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
NM_178080.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
NM_178080.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
NR_110360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019883.mm.1 |
TC0300002543.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
NM_001039376.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
NM_001039376.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
NM_001289701.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
NM_001289702.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
NM_178080.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
NM_178080.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
NR_110360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300019881.mm.1 |
TC0300002543.mm.1 |
MTA TC |