Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200018851.mm.1 |
NM_178111.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018851.mm.1 |
NM_178111.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018851.mm.1 |
TC0200002366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018853.mm.1 |
NM_178111.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018853.mm.1 |
NM_178111.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018853.mm.1 |
TC0200002366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018845.mm.1 |
NM_178111.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018845.mm.1 |
NM_178111.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018845.mm.1 |
TC0200002366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018855.mm.1 |
NM_178111.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018855.mm.1 |
NM_178111.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018855.mm.1 |
TC0200002366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018856.mm.1 |
NM_178111.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018856.mm.1 |
NM_178111.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018856.mm.1 |
TC0200002366.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018850.mm.1 |
NM_178111.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018850.mm.1 |
NM_178111.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018850.mm.1 |
TC0200002366.mm.1 |
MTA TC |