Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037136.1 | NP_001032213.1 | 4 - 5 | 1017 | 92 | |
NM_178119.3 | NP_835220.1 | 4 - 5 | 1017 | 92 | ||
GenBank mRNA | AK046523.1 | - | 4 - 5 | 978 | 92 | |
AK129289.2 | - | 4 - 5 | 964 | 92 | ||
AK147503.1 | - | 4 - 5 | 1023 | 92 | ||
AK147521.1 | - | 5 - 6 | 1668 | 93 | ||
BC096588.1 | - | 4 - 5 | 965 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |