Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178798.3 | NP_848913.1 | 1 - 2 | 82 | 105 | |
GenBank mRNA | AK030762.1 | - | 1 - 2 | 83 | 105 | |
AK046548.1 | - | 1 - 2 | 82 | 105 | ||
AK049478.1 | - | 1 - 2 | 82 | 105 | ||
AK053899.1 | - | 1 - 2 | 91 | 105 | ||
AK157379.1 | - | 1 - 2 | 83 | 105 | ||
AK164666.1 | - | 1 - 2 | 83 | 105 | ||
AK165629.1 | - | 1 - 2 | 99 | 105 | ||
AK165979.1 | - | 1 - 2 | 101 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |