Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178911.4 | NP_849242.1 | 7 - 8 | 1010 | 127 | |
GenBank mRNA | AK045459.1 | - | 7 - 8 | 969 | 127 | |
AK079549.1 | - | 7 - 8 | 969 | 127 | ||
AK079878.1 | - | 7 - 8 | 1012 | 127 | ||
AK151551.1 | - | 7 - 8 | 1244 | 127 | ||
AK152969.1 | - | 6 - 7 | 858 | 128 | ||
AK170362.1 | - | 7 - 8 | 967 | 127 | ||
BC058565.1 | - | 7 - 8 | 960 | 6442038 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |