Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178918.3 | NP_849249.1 | 7 - 8 | 1007 | 129 | |
GenBank mRNA | AK049186.1 | - | 7 - 8 | 1007 | 129 | |
AK162794.1 | - | 7 - 8 | 1011 | 129 | ||
AK165546.1 | - | 7 - 8 | 1002 | 129 | ||
AY151261.1 | - | 7 - 8 | 948 | 129 | ||
BC034372.1 | - | 3 - 4 | 364 | 4021387 | 129 | |
BC065057.1 | - | 7 - 8 | 1007 | 5715800 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |