Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007698.3 | NP_031724.2 | 1 - 2 | 460 | 83 | |
GenBank mRNA | AK081248.1 | - | 1 - 2 | 117 | 83 | |
AK138282.1 | - | 1 - 2 | 462 | 83 | ||
BC094242.1 | - | 1 - 2 | 444 | 5686074 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |