Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010786.4 | NP_034916.1 | 2 - 3 | 292 | 67 | |
GenBank mRNA | AK004719.1 | - | 2 - 3 | 251 | 67 | |
AK004881.1 | - | 2 - 3 | 219 | 67 | ||
AK088638.1 | - | 2 - 3 | 222 | 67 | ||
AK150530.1 | - | 1 - 2 | 80 | 67 | ||
AK152685.1 | - | 1 - 2 | 80 | 67 | ||
AK160069.1 | - | 1 - 2 | 83 | 67 | ||
AK168690.1 | - | 2 - 3 | 246 | 67 | ||
BC050902.1 | - | 2 - 3 | 208 | 6415937 | 67 | |
BC092270.1 | - | 2 - 3 | 245 | 4018419 | 67 | |
BY323121.1 | - | 2 - 3 | 293 | 67 | ||
X58876.1 | - | 2 - 3 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |