Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113418.1 | NP_001106889.1 | 3 - 4 | 418 | 86 | |
NM_011144.6 | NP_035274.2 | 3 - 4 | 656 | 86 | ||
GenBank mRNA | AK035676.1 | - | 3 - 4 | 657 | 86 | |
AK081709.1 | - | 3 - 4 | 419 | 86 | ||
AK149460.1 | - | 3 - 4 | 355 | 86 | ||
BC016892.1 | - | 3 - 4 | 649 | 4222384 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Nuclear Receptor Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |