Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011349.3 | NP_035479.2 | 5 - 6 | 656 | 67 | |
GenBank mRNA | AF036162.1 | - | 3 - 4 | 314 | 67 | |
AK053632.1 | - | 5 - 6 | 657 | 67 | ||
AK140340.1 | - | 5 - 6 | 1108 | 67 | ||
BC010976.1 | - | 5 - 6 | 577 | 4219060 | 67 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK172610.1 | - | 1049 | Chr. - - [-] on Build | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |