Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011401.4 | NP_035531.3 | 1 - 2 | 371 | 64 | |
GenBank mRNA | AK028041.1 | - | 1 - 2 | 145 | 64 | |
AK045328.1 | - | 1 - 2 | 147 | 64 | ||
AK147258.1 | - | 1 - 2 | 149 | 64 | ||
AK155997.1 | - | 1 - 2 | 130 | 64 | ||
AK164286.1 | - | 1 - 2 | 143 | 64 | ||
BC034122.1 | - | 1 - 2 | 129 | 4527667 | 64 | |
BC050095.1 | - | 1 - 2 | 127 | 64 | ||
BC058811.1 | - | 1 - 2 | 129 | 6490844 | 64 | |
BQ769655.1 | - | 1 - 2 | 373 | 64 | ||
M75135.1 | - | 1 - 2 | 95 | 64 | ||
X61093.1 | - | 1 - 2 | 95 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |