Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700036215.mm.1 |
NM_001285905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036215.mm.1 |
NM_001285906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036215.mm.1 |
NM_001285907.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036215.mm.1 |
NM_027353.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036215.mm.1 |
TC0700004315.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036221.mm.1 |
NM_001285905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036221.mm.1 |
NM_001285906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036221.mm.1 |
NM_001285907.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036221.mm.1 |
NM_027353.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036221.mm.1 |
TC0700004315.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700036223.mm.1 |
NM_001285905.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036223.mm.1 |
NM_001285906.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036223.mm.1 |
NM_001285907.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036223.mm.1 |
NM_027353.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700036223.mm.1 |
TC0700004315.mm.1 |
MTA TC |