Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tusc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030254.3 | NP_084530.3 | 899 | 101 | ||
GenBank mRNA | AK028182.1 | - | 875 | 101 | ||
AK088616.1 | - | 913 | 101 | |||
AK132766.1 | - | 922 | 101 | |||
BC069898.1 | - | 899 | 6489807 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |