Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100002374.mm.1 |
NM_053107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002374.mm.1 |
NM_053107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002374.mm.1 |
TC0100000317.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002367.mm.1 |
NM_053107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002367.mm.1 |
NM_053107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002367.mm.1 |
TC0100000317.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002369.mm.1 |
NM_053107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002369.mm.1 |
NM_053107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002369.mm.1 |
TC0100000317.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100002373.mm.1 |
NM_053107.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002373.mm.1 |
NM_053107.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100002373.mm.1 |
TC0100000317.mm.1 |
MTA TC |