Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297606.1 | NP_001284535.1 | 7 - 8 | 917 | 85 | |
NM_001297607.1 | NP_001284536.1 | 7 - 8 | 917 | 85 | ||
NM_009010.5 | NP_033036.2 | 7 - 8 | 914 | 85 | ||
GenBank mRNA | AK038300.1 | - | 7 - 8 | 891 | 85 | |
AK165467.1 | - | 7 - 8 | 918 | 85 | ||
BC003846.1 | - | 7 - 8 | 905 | 85 | ||
BC138684.1 | - | 7 - 8 | 874 | 8861705 | 85 | |
BC145372.1 | - | 7 - 8 | 877 | 9053930 | 85 | |
X92410.1 | - | 7 - 8 | 817 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |