Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030596.3 | NP_085099.2 | 7 - 8 | 881 | 134 | |
GenBank mRNA | AK029018.1 | - | 903 | 134 | ||
AK037142.1 | - | 7 - 8 | 912 | 134 | ||
AK148657.1 | - | 7 - 8 | 882 | 134 | ||
BC132058.1 | - | 7 - 8 | 910 | 40130335 | 134 | |
BC132639.1 | - | 7 - 8 | 910 | 40130916 | 134 | |
U86016.1 | - | 7 - 8 | 810 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |