Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195390.1 | NP_001182319.1 | 6 - 7 | 515 | 75 | |
NM_133850.2 | NP_598611.2 | 5 - 6 | 440 | 75 | ||
GenBank mRNA | AK008447.1 | - | 5 - 6 | 395 | 75 | |
AK009274.1 | - | 5 - 6 | 415 | 75 | ||
AK046582.1 | - | 5 - 6 | 390 | 75 | ||
AK077349.1 | - | 5 - 6 | 381 | 75 | ||
AK078774.1 | - | 5 - 6 | 399 | 75 | ||
AK089926.1 | - | 5 - 6 | 383 | 75 | ||
AK164805.1 | - | 5 - 6 | 349 | 75 | ||
AK169037.1 | - | 6 - 7 | 462 | 75 | ||
BC003310.1 | - | 5 - 6 | 373 | 75 | ||
BC065794.1 | - | 5 - 6 | 341 | 75 | ||
BC092215.1 | - | 5 - 6 | 357 | 6819383 | 75 | |
BC094216.1 | - | 5 - 6 | 397 | 6820900 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |