Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011394.3 | NP_035524.2 | 9 - 10 | 2204 | 53 | |
GenBank mRNA | AF196476.1 | - | 8 - 9 | 1972 | 53 | |
AK087507.1 | - | 9 - 10 | 2029 | 53 | ||
AK157206.1 | - | 9 - 10 | 2078 | 53 | ||
AK170985.1 | - | 9 - 10 | 2170 | 53 | ||
BC046510.1 | - | 9 - 10 | 2127 | 4035182 | 53 | |
BC094562.1 | - | 9 - 10 | 2584 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |