Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144748 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144742 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144747 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144743 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144745 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144746 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144741 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144744 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
390330 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
59647 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
681919 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
AB093249.2 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
AK005349.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
AK006863.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
AK037724.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
AK137407.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
AK139280.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
AK163484.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
BC079893.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
301393 |
NM_020252.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
796649 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
518935 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
771260 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
445591 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P125351 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P173267 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144758 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144751 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144749 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144753 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144737 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144756 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144741 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144764 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144743 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144738 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144736 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144760 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144735 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144732 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144733 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144747 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144745 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26144762 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P173267 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06631862 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P125351 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC117992.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC117993.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC054835.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
BC103793.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P266640 |
AK075755.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P117741 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P125221 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P125351 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P173267 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06926957 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06926962 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06926963 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06926967 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06926954 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P06926964 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144747 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144744 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144745 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144748 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144746 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144742 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144743 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26144741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144724 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144696 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144657 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144745 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144686 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144687 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144751 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144753 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144607 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144756 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144663 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144689 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144707 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144682 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26144619 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07024186 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07117562 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144722 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06786114 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06669814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06754346 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144756 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144714 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144733 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144767 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07112418 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144726 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144741 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144749 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07280489 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144732 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144721 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144762 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07013085 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144760 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144738 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144727 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144763 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144710 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07130519 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144729 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144745 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144709 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07383678 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07374888 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06763590 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144751 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144718 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144753 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P26144737 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P07130520 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06926518 |
- |
- |