Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026796.1 | NP_081072.1 | 5 - 6 | 545 | 86 | |
GenBank mRNA | AK145261.1 | - | 5 - 6 | 550 | 86 | |
AK150857.1 | - | 5 - 6 | 549 | 86 | ||
AK164053.1 | - | 6 - 7 | 1055 | 86 | ||
AK215563.1 | - | 1 - 2 | 75 | 86 | ||
BC023119.1 | - | 5 - 6 | 545 | 4458755 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |