Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026825.3 | NP_081101.3 | 31 - 32 | 3006 | 67 | |
GenBank mRNA | AK043591.1 | - | 17 - 18 | 1604 | 67 | |
AY437876.1 | - | 31 - 32 | 3011 | 67 | ||
BC012229.1 | - | 10 - 11 | 1035 | 67 | ||
BC150819.1 | - | 30 - 31 | 2920 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |