Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026825.3 | NP_081101.3 | 36 - 37 | 4041 | 70 | |
GenBank mRNA | AK043591.1 | - | 22 - 23 | 2639 | 70 | |
AY437876.1 | - | 36 - 37 | 4046 | 70 | ||
BC012229.1 | - | 15 - 16 | 2070 | 70 | ||
BC150819.1 | - | 35 - 36 | 3955 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |