Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026912.1 | NP_081188.1 | 4 - 5 | 438 | 86 | |
GenBank mRNA | BC016091.1 | - | 4 - 5 | 438 | 4503873 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |