Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027256.2 | NP_081532.1 | 15 - 16 | 2067 | 76 | |
GenBank mRNA | AK011676.1 | - | 1 - 2 | 64 | 76 | |
BC013710.1 | - | 15 - 16 | 2013 | 3156446 | 76 | |
BC013813.1 | - | 3 - 4 | 321 | 76 | ||
BC021838.1 | - | 8 - 9 | 1070 | 76 | ||
BC031145.1 | - | 9 - 10 | 1146 | 76 | ||
CX218921.1 | - | 4 - 5 | 429 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |