Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027427.2 | NP_081703.1 | 9 - 10 | 753 | 91 | |
GenBank mRNA | AK051568.1 | - | 9 - 10 | 754 | 91 | |
AK146331.1 | - | 9 - 10 | 716 | 91 | ||
AK166550.1 | - | 9 - 10 | 746 | 91 | ||
BC043716.1 | - | 3 - 4 | 81 | 91 | ||
BC137591.1 | - | 9 - 10 | 727 | 8860610 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |