Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111304.1 | NP_001104774.1 | 11 - 12 | 2644 | 72 | |
NM_027758.4 | NP_082034.1 | 8 - 9 | 1945 | 72 | ||
GenBank mRNA | AK049049.1 | - | 8 - 9 | 1946 | 72 | |
AK134614.1 | - | 11 - 12 | 1921 | 72 | ||
AK173050.1 | - | 11 - 12 | 1817 | 72 | ||
BC057027.1 | - | 7 - 8 | 1232 | 6825869 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |