Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zufsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028287.2 | NP_082563.1 | 1 - 2 | 82 | 79 | |
GenBank mRNA | AK012263.1 | - | 1 - 2 | 93 | 79 | |
AK046191.1 | - | 1 - 2 | 85 | 79 | ||
AK052071.1 | - | 1 - 2 | 66 | 79 | ||
AK077304.1 | - | 1 - 2 | 130 | 79 | ||
AK077730.1 | - | 1 - 2 | 103 | 79 | ||
AK160928.1 | - | 1 - 2 | 81 | 79 | ||
BC089514.1 | - | 1 - 2 | 95 | 30252639 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |